2,6-Difluoro-4-(methylsulfonyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

120001-500MG 120001-1G 120001-5G 120001-25G
77323-672EA 182.31 USD
77323-672 77347-092 77359-816 77383-618
2,6-Difluoro-4-(methylsulfonyl)benzoic acid ≥97%
2,6-Difluoro-4-(methylsulfonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₄S
MW: 236.193 g/mol
Storage Temperature: Ambient
CAS Number: 2918951-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR