1-(4-Bromo-3-iodophenyl)pyrrolidine ≥97%

Supplier: AOBChem USA

Warning

119910-500MG 119910-1G 119910-5G 119910-25G
77323-670EA 182.31 USD
77323-670 77347-090 77359-814 77383-616
1-(4-Bromo-3-iodophenyl)pyrrolidine ≥97%
1-(4-Bromo-3-iodophenyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrIN
MW: 352.01 g/mol
Storage Temperature: Ambient
CAS Number: 1696031-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR