2-Bromo-4-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

119831-1G 119831-5G 119831-25G
77341-898EA 106.09 USD
77341-898 77359-812 77371-808
2-Bromo-4-methoxybenzamide ≥97%
2-Bromo-4-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD01928225
CAS Number: 353278-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR