3,3'-dichloro-5,5'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

119803-500MG 119803-1G 119803-5G 119803-25G
77323-668EA 112.27 USD
77323-668 77343-338 77359-810 77371-806
3,3'-dichloro-5,5'-dimethyl-1,1'-biphenyl ≥97%
3,3'-dichloro-5,5'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂Cl₂
Storage Temperature: Ambient
CAS Number: 859080-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR