3’-Bromo-5’-fluoro-2’-hydroxyacetophenone ≥95%

Supplier: AOBChem USA

Warning

119768-500MG 119768-1G 119768-5G 119768-25G
77323-662EA 211.15 USD
77323-662 77347-744 77359-804 77383-610
3’-Bromo-5’-fluoro-2’-hydroxyacetophenone ≥95%
3’-Bromo-5’-fluoro-2’-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD16742831
CAS Number: 393-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR