5-Chloro-3-fluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

119674-500MG 119674-1G 119674-5G
77339-888EA 671.56 USD
77339-888 77347-742 77371-804
5-Chloro-3-fluoro-2-methylbenzaldehyde ≥97%
5-Chloro-3-fluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD28751758
CAS Number: 1805225-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR