5-Chloro-3-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

119672-500MG 119672-1G 119672-5G
77323-660EA 350.2 USD
77323-660 77347-740 77371-802
5-Chloro-3-fluoro-2-methylbenzoic acid ≥97%
5-Chloro-3-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21602115
CAS Number: 1365969-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR