4-Methyl-5-(methylthio)benzo[d][1,3]dioxole ≥97%

Supplier: AOBChem USA

Warning

119631-500MG 119631-1G 119631-5G
77340-356EA 919.79 USD
77340-356 77347-736 77371-798
4-Methyl-5-(methylthio)benzo[d][1,3]dioxole ≥97%
4-Methyl-5-(methylthio)benzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₉H₁₀O₂S
Storage Temperature: Ambient
CAS Number: 70080-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR