1-(3-Chloro-2-fluoro-4-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

119599-500MG 119599-1G 119599-5G
77335-656EA 361.53 USD
77335-656 77347-734 77371-796
1-(3-Chloro-2-fluoro-4-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-Chloro-2-fluoro-4-methoxyphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅ClF₄O₂
Storage Temperature: Ambient
CAS Number: 2921693-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR