5-(2-Bromophenoxy)-2-nitrobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

119298-500MG 119298-1G 119298-5G 119298-25G
77323-656EA 88.58 USD
77323-656 77342-342 77359-800 77371-794
5-(2-Bromophenoxy)-2-nitrobenzoic acid ≥97%
5-(2-Bromophenoxy)-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₈BrNO₅
Storage Temperature: Ambient
MDL Number: MFCD09933214
CAS Number: 1016778-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR