3-(1,1-Difluoroethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

119116-500MG 119116-1G 119116-5G
77323-652EA 316.21 USD
77323-652 77347-732 77371-782
3-(1,1-Difluoroethyl)benzaldehyde ≥97%
3-(1,1-Difluoroethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O
MW: 170.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20527725
CAS Number: 1032527-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR