2-(1,1-Difluoroethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

119112-500MG 119112-1G 119112-5G 119112-25G
77323-650EA 173.04 USD
77323-650 77346-468 77359-790 77383-606
2-(1,1-Difluoroethyl)benzaldehyde ≥97%
2-(1,1-Difluoroethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O
MW: 170.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28519105
CAS Number: 1782543-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR