1-Fluoro-3-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

119090-500MG 119090-1G 119090-25G 119090-5G
77323-648EA 300.76 USD
77323-648 77347-728 77383-604 77371-778
1-Fluoro-3-(methoxymethyl)benzene ≥95%
1-Fluoro-3-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₉FO
Storage Temperature: Ambient
MDL Number: MFCD04116346
CAS Number: 28490-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR