3-Cyclopentyl-4-fluoro-5-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

118969-500MG 118969-1G
77340-562EA 1902.41 USD
77340-562 77347-726
3-Cyclopentyl-4-fluoro-5-methylbenzaldehyde ≥95%
3-Cyclopentyl-4-fluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD34784270
CAS Number: 2901106-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR