3-Cyclopentyl-4-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

118967-500MG 118967-1G
77340-456EA 1123.73 USD
77340-456 77347-724
3-Cyclopentyl-4-fluoro-5-methylbenzoic acid ≥97%
3-Cyclopentyl-4-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅FO₂
MW: 222.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784268
CAS Number: 2901097-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR