3-Bromo-4-fluorophenylacetylene ≥95%

Supplier: AOBChem USA

Warning

118596-500MG 118596-1G 118596-5G 118596-25G
77323-646EA 172.01 USD
77323-646 77346-428 77359-788 77383-602
3-Bromo-4-fluorophenylacetylene ≥95%
3-Bromo-4-fluorophenylacetylene

Precaución: for Research Use Only

Formula: C₈H₄BrF
Storage Temperature: Ambient
MDL Number: MFCD26395447
CAS Number: 918487-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR