(2-Chloro-5-fluorophenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118491-500MG 118491-1G 118491-5G 118491-25G
77323-642EA 161.71 USD
77323-642 77346-054 77359-780 77383-600
(2-Chloro-5-fluorophenyl)(phenyl)methanol ≥97%
(2-Chloro-5-fluorophenyl)(phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClFO
MW: 236.669 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784115
CAS Number: 1594502-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR