(2-Methoxyphenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118436-500MG 118436-1G 118436-5G
77337-666EA 434.66 USD
77337-666 77347-720 77371-766
(2-Methoxyphenyl)(phenyl)methanol ≥97%
(2-Methoxyphenyl)(phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06654248
CAS Number: 22788-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR