(2-Chlorophenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118434-1G 118434-5G 118434-25G
77341-492EA 89.61 USD
77341-492 77359-778 77371-764
(2-Chlorophenyl)(phenyl)methanol ≥97%
(2-Chlorophenyl)(phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD00030010
CAS Number: 6954-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR