2-(Trifluoromethyl)benzhydrol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Phenyl(2-(trifluoromethyl)phenyl)methanol

118422-1G 118422-5G 118422-25G
77340-862EA 60.77 USD
77340-862 77359-776 77371-762
2-(Trifluoromethyl)benzhydrol ≥97%
2-(Trifluoromethyl)benzhydrol

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₃O
MW: 252.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014396
CAS Number: 727-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR