2-Fluoro-5-methylisophthalaldehyde ≥97%

Supplier: AOBChem USA

Warning

118384-500MG 118384-1G 118384-5G 118384-25G
77323-634EA 82.4 USD
77323-634 77342-174 77359-770 77371-756
2-Fluoro-5-methylisophthalaldehyde ≥97%
2-Fluoro-5-methylisophthalaldehyde

Precaución: for Research Use Only

Formula: C₉H₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD34784092
CAS Number: 2091398-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR