4-Methylbenzo[d][1,3]dioxol-5-ol ≥97%

Supplier: AOBChem USA

Warning

118369-500MG 118369-1G 118369-5G
77335-922EA 367.71 USD
77335-922 77347-714 77371-754
4-Methylbenzo[d][1,3]dioxol-5-ol ≥97%
4-Methylbenzo[d][1,3]dioxol-5-ol

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18805454
CAS Number: 187040-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR