(6-Bromo-2-fluoro-3-(methylthio)phenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118186-1G 118186-5G 118186-25G
77342-068EA 111.24 USD
77342-068 77359-766 77371-752
(6-Bromo-2-fluoro-3-(methylthio)phenyl)(phenyl)methanol ≥97%
(6-Bromo-2-fluoro-3-(methylthio)phenyl)(phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFOS
MW: 327.212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783942
CAS Number: 2901085-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR