(2-Bromo-6-fluoro-3-propoxyphenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118164-1G 118164-5G 118164-25G
77341-728EA 101.97 USD
77341-728 77359-762 77371-748
(2-Bromo-6-fluoro-3-propoxyphenyl)(phenyl)methanol ≥97%
(2-Bromo-6-fluoro-3-propoxyphenyl)(phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₆H₁₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34783920
CAS Number: 2901105-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR