4-Fluoro-3-(1-hydroxy-2-methylpropyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

118114-500MG 118114-1G 118114-5G 118114-25G
77323-630EA 280.16 USD
77323-630 77347-712 77359-758 77383-592
4-Fluoro-3-(1-hydroxy-2-methylpropyl)benzonitrile ≥97%
4-Fluoro-3-(1-hydroxy-2-methylpropyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO
MW: 193.217 g/mol
Storage Temperature: Ambient
CAS Number: 1508870-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR