(3-Chloro-5-(cyclopropylcarbamoyl)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

118010-500MG 118010-1G 118010-5G 118010-25G
77323-628EA 202.91 USD
77323-628 77347-710 77359-756 77383-590
(3-Chloro-5-(cyclopropylcarbamoyl)phenyl)boronic acid ≥97%
(3-Chloro-5-(cyclopropylcarbamoyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD28167462
CAS Number: 1423127-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR