2-Bromo-3-fluoro-4-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

117945-500MG 117945-1G 117945-5G
77338-586EA 501.61 USD
77338-586 77347-708 77371-744
2-Bromo-3-fluoro-4-methoxyphenol ≥97%
2-Bromo-3-fluoro-4-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28738840
CAS Number: 1600530-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR