6-(Cyclopent-1-en-1-yl)-2,3-dihydrobenzo[b][1,4]dioxine ≥97%

Supplier: AOBChem USA

Warning

117922-500MG 117922-1G 117922-25G 117922-5G
77323-626EA 297.67 USD
77323-626 77347-706 77383-588 77371-740
6-(Cyclopent-1-en-1-yl)-2,3-dihydrobenzo[b][1,4]dioxine ≥97%
6-(Cyclopent-1-en-1-yl)-2,3-dihydrobenzo[b][1,4]dioxine

Precaución: for Research Use Only

Formula: C₁₃H₁₄O₂
MW: 202.2490 g/mol
Storage Temperature: Ambient
CAS Number: 2918931-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR