4-Bromo-N,N,3-trimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

11792-1G 11792-5G 11792-25G
77340-982EA 66.95 USD
77340-982 77359-754 77371-742
4-Bromo-N,N,3-trimethylbenzamide ≥97%
4-Bromo-N,N,3-trimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO
Storage Temperature: Ambient
MDL Number: MFCD26395567
CAS Number: 149104-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR