2-(methoxymethoxy)-3-(trimethylsilyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

117791-500MG 117791-1G 117791-5G 117791-25G
77323-624EA 105.06 USD
77323-624 77343-082 77359-752 77371-738
2-(methoxymethoxy)-3-(trimethylsilyl)benzaldehyde ≥97%
2-(methoxymethoxy)-3-(trimethylsilyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₃Si
MW: 238.355 g/mol
Storage Temperature: Ambient
CAS Number: 140840-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR