1-(Cyclopent-1-en-1-yl)-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

117773-500MG 117773-1G 117773-5G
77337-716EA 437.75 USD
77337-716 77347-704 77371-736
1-(Cyclopent-1-en-1-yl)-2-isopropoxybenzene ≥97%
1-(Cyclopent-1-en-1-yl)-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₈O
MW: 202.2921 g/mol
Storage Temperature: Ambient
CAS Number: 2901081-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR