2-Chloro-1-(3-Chloro-4-methylphenyl)-2,2-difluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
117751-500MG
117751-1G
117751-5G
117751-25G
77323-622EA
220.42
USD
77323-622
77347-702
77359-750
77383-586
2-Chloro-1-(3-Chloro-4-methylphenyl)-2,2-difluoroethanone ≥97%
2-Chloro-1-(3-Chloro-4-methylphenyl)-2,2-difluoroethanone
Precaución: for Research Use Only
Formula:
C₉H₆Cl₂F₂O MW: 239.0461 g/mol Storage Temperature: Ambient |
CAS Number:
1352231-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...