2-Chloro-1-(3,5-dichlorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

117750-500MG 117750-1G 117750-5G 117750-25G
77323-620EA 220.42 USD
77323-620 77347-700 77359-748 77383-584
2-Chloro-1-(3,5-dichlorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(3,5-dichlorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₃Cl₃F₂O
MW: 259.4646 g/mol
Storage Temperature: Ambient
CAS Number: 864725-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR