2-Chloro-2,2-difluoro-1-(4-(trifluoromethyl)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

117749-500MG 117749-1G 117749-5G
77336-968EA 400.67 USD
77336-968 77347-698 77371-734
2-Chloro-2,2-difluoro-1-(4-(trifluoromethyl)phenyl)ethanone ≥97%
2-Chloro-2,2-difluoro-1-(4-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₄ClF₅O
MW: 258.5724 g/mol
Storage Temperature: Ambient
CAS Number: 1321028-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR