2-Chloro-2,2-difluoro-1-(2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

117744-500MG 117744-1G 117744-5G 117744-25G
77323-612EA 223.51 USD
77323-612 77347-690 77359-742 77383-576
2-Chloro-2,2-difluoro-1-(2-fluorophenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-fluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
MW: 208.5649 g/mol
Storage Temperature: Ambient
CAS Number: 1566680-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR