2,8-Dichloro-6-methylquinoline ≥97%

Supplier: AOBChem USA

Warning

117580-500MG 117580-1G 117580-5G
77337-998EA 456.29 USD
77337-998 77347-684 77371-722
2,8-Dichloro-6-methylquinoline ≥97%
2,8-Dichloro-6-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₇Cl₂N
MW: 212.0752 g/mol
Storage Temperature: Ambient
CAS Number: 1195523-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR