1-(3-(benzyloxy)-2-bromo-6-fluorophenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

117440-1G 117440-5G 117440-25G
77340-860EA 60.77 USD
77340-860 77359-732 77371-718
1-(3-(benzyloxy)-2-bromo-6-fluorophenyl)-2-methylpropan-1-ol ≥95%
1-(3-(benzyloxy)-2-bromo-6-fluorophenyl)-2-methylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₇H₁₈BrFO₂
MW: 353.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783476
CAS Number: 2901109-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR