1-(6-Bromo-2-fluoro-3-methylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

117432-500MG 117432-1G 117432-25G 117432-5G
77323-604EA 305.91 USD
77323-604 77347-680 77383-568 77371-716
1-(6-Bromo-2-fluoro-3-methylphenyl)-2-methylpropan-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-methylphenyl)-2-methylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrFO
Storage Temperature: Ambient
MDL Number: MFCD34783468
CAS Number: 1503458-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR