4-Fluoro-2-methoxy-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

117265-500MG 117265-1G 117265-5G 117265-25G
77323-602EA 117.42 USD
77323-602 77343-520 77359-730 77371-710
4-Fluoro-2-methoxy-5-methylphenol ≥97%
4-Fluoro-2-methoxy-5-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23110601
CAS Number: 1395035-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR