1-Bromo-2,4-dimethyl-5-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

117260-1G 117260-5G 117260-25G
77341-748EA 103 USD
77341-748 77359-728 77371-708
1-Bromo-2,4-dimethyl-5-(methylsulfonyl)benzene ≥97%
1-Bromo-2,4-dimethyl-5-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂S
MW: 263.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783429
CAS Number: 141763-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR