2-Iodo-m-toluidine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Iodo-3-methylaniline

117240-1G 117240-5G 117240-25G
77341-064EA 67.98 USD
77341-064 77359-724 77371-706
2-Iodo-m-toluidine ≥97%
2-Iodo-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈IN
MW: 233.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505640
CAS Number: 89938-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR