3-Bromo-5-fluoro-2-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

116902-500MG 116902-1G 116902-5G
77335-598EA 359.47 USD
77335-598 77347-668 77371-698
3-Bromo-5-fluoro-2-isopropoxybenzaldehyde ≥97%
3-Bromo-5-fluoro-2-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28396359
CAS Number: 1781579-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR