Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate

116899-500MG 116899-1G 116899-5G 116899-25G
77323-598EA 132.87 USD
77323-598 77344-408 77359-718 77383-564
Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate ≥97%
Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01546256
CAS Number: 214894-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR