3-(4-cyanophenoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

116717-500MG 116717-1G 116717-5G 116717-25G
77323-596EA 184.37 USD
77323-596 77347-178 77359-716 77383-562
3-(4-cyanophenoxy)benzoic acid ≥95%
3-(4-cyanophenoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00447014
CAS Number: 313549-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR