1,1,1-trifluoro-2-(4-fluoro-2-methylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

116711-500MG 116711-1G 116711-5G 116711-25G
77323-590EA 182.31 USD
77323-590 77347-084 77359-710 77383-556
1,1,1-trifluoro-2-(4-fluoro-2-methylphenyl)propan-2-ol ≥97%
1,1,1-trifluoro-2-(4-fluoro-2-methylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₄O
Storage Temperature: Ambient
CAS Number: 2148819-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR