2-(3-Bromophenyl)-1,1,1-trifluoropropan-2-ol ≥97%

Supplier: AOBChem USA

Warning

116709-500MG 116709-1G 116709-5G
77323-588EA 329.6 USD
77323-588 77347-664 77371-694
2-(3-Bromophenyl)-1,1,1-trifluoropropan-2-ol ≥97%
2-(3-Bromophenyl)-1,1,1-trifluoropropan-2-ol

Precaución: for Research Use Only

Formula: C₉H₈BrF₃O
MW: 269.0584 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16068003
CAS Number: 122243-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR