2-(2,4-Difluorophenyl)-1,1,1-trifluoropropan-2-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
116707-500MG
116707-1G
116707-5G
116707-25G
77323-586EA
182.31
USD
77323-586
77347-082
77359-708
77383-554
2-(2,4-Difluorophenyl)-1,1,1-trifluoropropan-2-ol ≥97%
2-(2,4-Difluorophenyl)-1,1,1-trifluoropropan-2-ol
Precaución: for Research Use Only
Formula:
C₉H₇F₅O MW: 226.143 g/mol Storage Temperature: Ambient |
CAS Number:
1248360-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...