(2-Fluorophenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

116706-500MG 116706-1G 116706-5G
77336-002EA 372.86 USD
77336-002 77347-662 77371-692
(2-Fluorophenyl)(phenyl)methanol ≥97%
(2-Fluorophenyl)(phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₁FO
Storage Temperature: Ambient
CAS Number: 362-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR