1,1,1-trifluoro-2-(2-fluorophenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

116705-500MG 116705-1G 116705-5G 116705-25G
77323-584EA 182.31 USD
77323-584 77347-080 77359-706 77383-552
1,1,1-trifluoro-2-(2-fluorophenyl)propan-2-ol ≥97%
1,1,1-trifluoro-2-(2-fluorophenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₉H₈F₄O
MW: 208.1528 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16067977
CAS Number: 122243-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR