1,1,1-trifluoro-2-(4-fluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

116701-500MG 116701-1G 116701-5G
77337-784EA 442.9 USD
77337-784 77347-660 77371-690
1,1,1-trifluoro-2-(4-fluorophenyl)propan-2-ol ≥95%
1,1,1-trifluoro-2-(4-fluorophenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₉H₈F₄O
Storage Temperature: Ambient
CAS Number: 711-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR